2-Fluoro-3-(4-methylphenyl)-3-oxopropanenitrile 95%
Supplier: Apollo Scientific
PC28205-1G
PC28205-5G
APOSPC28205-1GEA
234
EUR
APOSPC28205-1G
APOSPC28205-5G
2-Fluoro-3-(4-methylphenyl)-3-oxopropanenitrile 95%
2-Fluoro-3-(4-methylphenyl)-3-oxopropanenitrile
Formula:
C₁₀H₈FNO Storage Temperature: Ambient |
MDL Number:
MFCD18426512 CAS Number: 1263063-10-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...