(±)-3,5-Difluoromandelic acid
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3,5-Difluoromandelic acid, (±)-3,5-Difluorophenylglycolic acid
PC2842G-5G
APOSPC2842G-5GEA
66.7
EUR
APOSPC2842G-5G
(±)-3,5-Difluoromandelic acid
(±)-3,5-Difluoromandelic acid
Formula:
C₈H₆F₂O₃ MW: 188,13 g/mol Melting Pt: 135…139 °C Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00061300 CAS Number: 132741-31-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...