Hexafluoro-2-methylisopropyl acrylate 98%

Supplier: Apollo Scientific
Warning

Synonyms: 1,1,1,3,3,3-Hexafluoro-2-methylisopropyl acrylate

PC2843-1G PC2843-5G
APOSPC2843-1GEA 105 EUR
APOSPC2843-1G APOSPC2843-5G
Hexafluoro-2-methylisopropyl acrylate 98%
Hexafluoro-2-methylisopropyl acrylate
Formula: C₇H₆F₆O₂
MW: 236,12 g/mol
Boiling Pt: 60…61 °C/190mmHg
Storage Temperature: Ambient
MDL Number: MFCD03094206
CAS Number: 53038-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR