cis-1,2,3,3,3-Pentafluoropropene 97%
Supplier: Apollo Scientific
|
Warning
|
PC2923-25G
PC2923-5G
PC2923-100G
PC2923-500G
APOSPC2923-25GEA
270
EUR
APOSPC2923-25G
APOSPC2923-5G
APOSPC2923-100G
APOSPC2923-500G
cis-1,2,3,3,3-Pentafluoropropene 97%
cis-1,2,3,3,3-Pentafluoropropene
Formula:
C₃HF₅ MW: 132,03 g/mol Boiling Pt: –18 °C Storage Temperature: Ambient |
CAS Number:
5528-43-8 EINECS: 226-875-0 UN: 3163 ADR: 2.2, |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...