3-(Trifluoromethyl)benzenesulphonamide

Supplier: Apollo Scientific
Warning

Synonyms: α,α,α-Trifluoro-m-toluenesulphonamide

PC2979-10G PC2979-1G PC2979-5G
APOSPC2979-10GEA 114 EUR
APOSPC2979-10G APOSPC2979-1G APOSPC2979-5G
3-(Trifluoromethyl)benzenesulphonamide
3-(Trifluoromethyl)benzenesulphonamide
Formula: C₇H₆F₃NO₂S
Melting Pt: 122…126 °C
Storage Temperature: Ambient
MDL Number: MFCD00042421
CAS Number: 672-58-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR