5-(Trifluoromethoxy)indole-2-carboxylic acid 98%

Supplier: Apollo Scientific
Warning

Synonyms: 5-(Trifluoromethoxy)-1H-indole-2-carboxylic acid

PC3003-1G PC3003-5G
APOSPC3003-1GEA 116 EUR
APOSPC3003-1G APOSPC3003-5G
5-(Trifluoromethoxy)indole-2-carboxylic acid 98%
5-(Trifluoromethoxy)indole-2-carboxylic acid
Formula: C₁₀H₆F₃NO₃
Melting Pt: 201…207 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00276997
CAS Number: 175203-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR