2-Fluoro-5-formylphenylboronic acid

Supplier: Apollo Scientific
Warning

Synonyms: 2-Fluoro-5-formylbenzeneboronic acid

PC3009-1G PC3009-25G PC3009-5G
APOSPC3009-1GEA 21.9 EUR
APOSPC3009-1G APOSPC3009-25G APOSPC3009-5G
2-Fluoro-5-formylphenylboronic acid
2-Fluoro-5-formylphenylboronic acid
Formula: C₇H₆BFO₃
MW: 167,93 g/mol
Melting Pt: 168…171 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03095100
CAS Number: 352534-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR