3-Fluoro-4-biphenylboronic acid

Supplier: Apollo Scientific
Warning

PC3012-1G PC3012-250MG
APOSPC3012-1GEA 383 EUR
APOSPC3012-1G APOSPC3012-250MG
3-Fluoro-4-biphenylboronic acid
3-Fluoro-4-biphenylboronic acid
Formula: C₁₂H₁₀BFO₂
Melting Pt: 237…242 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04115668
CAS Number: 409108-13-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR