3,4-Difluorobenzamidoxime
Supplier: Apollo Scientific
PC302036-10G
PC302036-50G
APOSPC302036-10GEA
79.9
EUR
APOSPC302036-10G
APOSPC302036-50G
3,4-Difluorobenzamidoxime
3,4-Difluorobenzamidoxime
Formula:
C₇H₆F₂N₂O MW: 172.13 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06660194 CAS Number: 500024-74-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...