2-Fluoro-4-methylbenzylamine 98%
Supplier: Apollo Scientific
|
Danger
|
PC302075-1G
PC302075-5G
APOSPC302075-1GEA
263
EUR
APOSPC302075-1G
APOSPC302075-5G
2-Fluoro-4-methylbenzylamine 98%
2-Fluoro-4-methylbenzylamine
Formula:
C₈H₁₀FN MW: 139,17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06213635 CAS Number: 771573-01-4 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...