(3-Chloro-2,4-difluorophenyl)acetic acid 97%
Supplier: Apollo Scientific
Synonyms:
3-Chloro-2,4-difluorophenylacetic acid
PC302087-1G
PC302087-5G
PC302087-250MG
APOSPC302087-1GEA
87
EUR
APOSPC302087-1G
APOSPC302087-5G
APOSPC302087-250MG
(3-Chloro-2,4-difluorophenyl)acetic acid 97%
(3-Chloro-2,4-difluorophenyl)acetic acid
Formula:
C₈H₅ClF₂O₂ MW: 206,58 g/mol Melting Pt: 124…126 °C Storage Temperature: Ambient |
MDL Number:
MFCD06660337 CAS Number: 886761-66-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...