2,6-Difluoro-4-methoxyphenylacetonitrile 98%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
2-(2,6-difluoro-4-methoxyphenyl)ethanenitrile, 2-(2,6-difluoro-4-methoxyphenyl)acetonitrile
PC302164-1G
PC302164-5G
PC302164-25G
APOSPC302164-1GEA
102
EUR
APOSPC302164-1G
APOSPC302164-5G
APOSPC302164-25G
2,6-Difluoro-4-methoxyphenylacetonitrile 98%
2,6-Difluoro-4-methoxyphenylacetonitrile
Formula:
C₉H₇F₂NO Storage Temperature: Ambient |
MDL Number:
MFCD04115928 CAS Number: 886499-03-2 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...