2-Methoxy-5-(trifluoromethoxy)benzonitrile 97%
Supplier: Apollo Scientific
PC302176-5G
PC302176-1G
PC302176-25G
APOSPC302176-5GEA
89.9
EUR
APOSPC302176-5G
APOSPC302176-1G
APOSPC302176-25G
2-Methoxy-5-(trifluoromethoxy)benzonitrile 97%
2-Methoxy-5-(trifluoromethoxy)benzonitrile
Formula:
C₉H₆F₃NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD04115963 CAS Number: 886500-03-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...