2-Chloro-6-fluoro-3-methoxybenzoic acid 97%

Supplier: Apollo Scientific
Warning

PC302233-25G PC302233-5G PC302233-1G
APOSPC302233-25GEA 739 EUR
APOSPC302233-25G APOSPC302233-5G APOSPC302233-1G
2-Chloro-6-fluoro-3-methoxybenzoic acid 97%
2-Chloro-6-fluoro-3-methoxybenzoic acid
Formula: C₈H₆ClFO₃
MW: 204,59 g/mol
Melting Pt: 126…128 °C
Storage Temperature: Ambient
MDL Number: MFCD04115942
CAS Number: 886499-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR