3,5-Difluoro-2-methylbenzaldehyde 98%

Supplier: Apollo Scientific
Warning

Synonyms: 3,5-Difluoro-o-tolylaldehyde, 3,5-Difluoro-2-tolylaldehyde, 3,5-Difluoro-2-tolualdehyde, 3,5-Difluoro-o-tolualdehyde

PC302271-1G PC302271-5G PC302271-10G
APOSPC302271-1GEA 150 EUR
APOSPC302271-1G APOSPC302271-5G APOSPC302271-10G
3,5-Difluoro-2-methylbenzaldehyde 98%
3,5-Difluoro-2-methylbenzaldehyde
Formula: C₈H₆F₂O
MW: 156,13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09832335
CAS Number: 1017778-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR