5-Methyl-2-(trifluoromethyl)benzyl bromide 98%

Supplier: Apollo Scientific

PC302362-1G PC302362-5G
APOSPC302362-1GEA 92.8 EUR
APOSPC302362-1G APOSPC302362-5G
5-Methyl-2-(trifluoromethyl)benzyl bromide 98%
5-Methyl-2-(trifluoromethyl)benzyl bromide
Formula: C₉H₈BrF₃
MW: 253,06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660296
CAS Number: 886502-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR