5-Methyl-2-(trifluoromethyl)benzylamine 98%
Supplier: Apollo Scientific
|
Danger
|
PC302363-1G
PC302363-5G
PC302363-250MG
APOSPC302363-1GEA
335
EUR
APOSPC302363-1G
APOSPC302363-5G
APOSPC302363-250MG
5-Methyl-2-(trifluoromethyl)benzylamine 98%
5-Methyl-2-(trifluoromethyl)benzylamine
Formula:
C₉H₁₀F₃N MW: 189,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09832358 CAS Number: 1017779-30-4 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...