6-Chloro-2-fluoro-3-methoxybenzoic acid 97%
Supplier: Apollo Scientific
PC302368-25G
PC302368-5G
PC302368-1G
APOSPC302368-25GEA
416
EUR
APOSPC302368-25G
APOSPC302368-5G
APOSPC302368-1G
6-Chloro-2-fluoro-3-methoxybenzoic acid 97%
6-Chloro-2-fluoro-3-methoxybenzoic acid
Formula:
C₈H₆ClFO₃ MW: 204,59 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04115947 CAS Number: 886499-58-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...