3-Fluoro-5-(trifluoromethoxy)benzyl bromide 98%
Supplier: Apollo Scientific
|
Danger
|
PC302554-1G
PC302554-5G
PC302554-10G
APOSPC302554-1GEA
609
EUR
APOSPC302554-1G
APOSPC302554-5G
APOSPC302554-10G
3-Fluoro-5-(trifluoromethoxy)benzyl bromide 98%
3-Fluoro-5-(trifluoromethoxy)benzyl bromide
Formula:
C₈H₅BrF₄O Storage Temperature: Ambient |
MDL Number:
MFCD18783226 CAS Number: 86256-51-1 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...