(4-Methoxy-2-(trifluoromethyl)phenyl)acetic acid 98%
Supplier: Apollo Scientific
|
Warning
|
PC302601-1G
PC302601-5G
PC302601-10G
APOSPC302601-1GEA
120
EUR
APOSPC302601-1G
APOSPC302601-5G
APOSPC302601-10G
(4-Methoxy-2-(trifluoromethyl)phenyl)acetic acid 98%
(4-Methoxy-2-(trifluoromethyl)phenyl)acetic acid
Formula:
C₁₀H₉F₃O₃ Melting Pt: 96…99 °C Storage Temperature: Ambient |
CAS Number:
916420-90-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...