6-Chloro-2-fluoro-3-methylbenzonitrile 98%

Supplier: Apollo Scientific
Danger

PC302635-25G PC302635-5G PC302635-1G PC302635-10G
APOSPC302635-25GEA 0 EUR
APOSPC302635-25G APOSPC302635-5G APOSPC302635-1G APOSPC302635-10G
6-Chloro-2-fluoro-3-methylbenzonitrile 98%
6-Chloro-2-fluoro-3-methylbenzonitrile
Formula: C₈H₅ClFN
MW: 169,59 g/mol
Melting Pt: 56…58 °C
Storage Temperature: Ambient
MDL Number: MFCD06660186
CAS Number: 886502-19-8
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR