6-Chloro-2-fluoro-3-methylaniline 98%

Supplier: Apollo Scientific

PC302651-10G PC302651-5G PC302651-1G
APOSPC302651-10GEA 224 EUR
APOSPC302651-10G APOSPC302651-5G APOSPC302651-1G
6-Chloro-2-fluoro-3-methylaniline 98%
6-Chloro-2-fluoro-3-methylaniline
Formula: C₇H₇ClFN
MW: 159,59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660187
CAS Number: 702640-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR