2-Chloro-5-(pentafluorothio)benzoyl chloride 97%
Supplier: Apollo Scientific
Synonyms:
2-Chloro-5-(pentafluorosulphur)benzoyl chloride
PC302674-1G
PC302674-5G
PC302674-10G
APOSPC302674-1GEA
350
EUR
APOSPC302674-1G
APOSPC302674-5G
APOSPC302674-10G
2-Chloro-5-(pentafluorothio)benzoyl chloride 97%
2-Chloro-5-(pentafluorothio)benzoyl chloride
Formula:
C₇H₃Cl₂F₅OS MW: 301,06 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22988932 CAS Number: 1431329-83-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...