3,6-Dichloro-2-fluorobenzonitrile 98%

Supplier: Apollo Scientific

PC302702-25G PC302702-5G PC302702-1G
APOSPC302702-25GEA 468 EUR
APOSPC302702-25G APOSPC302702-5G APOSPC302702-1G
3,6-Dichloro-2-fluorobenzonitrile 98%
3,6-Dichloro-2-fluorobenzonitrile
Formula: C₇H₂Cl₂FN
MW: 190,00 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09025385
CAS Number: 916420-65-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR