3'-Ethoxy-2',4'-difluoroacetophenone 98%

Supplier: Apollo Scientific

Synonyms: 2,4-Difluoro-3-ethoxy acetophenone

PC302720-10G PC302720-5G PC302720-1G
APOSPC302720-10GEA 282 EUR
APOSPC302720-10G APOSPC302720-5G APOSPC302720-1G
3'-Ethoxy-2',4'-difluoroacetophenone 98%
3'-Ethoxy-2',4'-difluoroacetophenone
Formula: C₁₀H₁₀F₂O₂
MW: 200,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09258734
CAS Number: 1017778-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR