5-Chloro-2-(trifluoromethoxy)aniline 97%

Supplier: Apollo Scientific

PC302758-5G PC302758-10G
APOSPC302758-5GEA 14.7 EUR
APOSPC302758-5G APOSPC302758-10G
5-Chloro-2-(trifluoromethoxy)aniline 97%
5-Chloro-2-(trifluoromethoxy)aniline
Formula: C₇H₅ClF₃NO
MW: 211,57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06410919
CAS Number: 326-64-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR