4-Ethoxy-3-(trifluoromethyl)benzyl alcohol 97%
Supplier: Apollo Scientific
Synonyms:
4-ETHOXY-3-(TRIFLUOROMETHYL)BENZYL ALCOHOL
PC302873-10G
PC302873-5G
APOSPC302873-10GEA
396
EUR
APOSPC302873-10G
APOSPC302873-5G
4-Ethoxy-3-(trifluoromethyl)benzyl alcohol 97%
4-Ethoxy-3-(trifluoromethyl)benzyl alcohol
Formula:
C₁₀H₁₀F₃NO₂ MW: 233,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13196581 CAS Number: 1206593-33-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...