5-Methyl-2-(trifluoromethyl)benzaldehyde 98%

Supplier: Apollo Scientific

PC302893-5G PC302893-1G
APOSPC302893-5GEA 147 EUR
APOSPC302893-5G APOSPC302893-1G
5-Methyl-2-(trifluoromethyl)benzaldehyde 98%
5-Methyl-2-(trifluoromethyl)benzaldehyde
Formula: C₉H₇F₃O
MW: 188,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660294
CAS Number: 886502-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR