5-Methyl-2-(trifluoromethyl)benzoic acid 98%

Supplier: Apollo Scientific
Warning

PC302894-10G PC302894-5G PC302894-1G
APOSPC302894-10GEA 282 EUR
APOSPC302894-10G APOSPC302894-5G APOSPC302894-1G
5-Methyl-2-(trifluoromethyl)benzoic acid 98%
5-Methyl-2-(trifluoromethyl)benzoic acid
Formula: C₉H₇F₃O₂
MW: 204,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660291
CAS Number: 120985-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR