6-Chloro-2-fluoro-3-methoxybenzylamine 97%
Supplier: Apollo Scientific
|
Danger
|
PC302899-10G
PC302899-5G
PC302899-1G
APOSPC302899-10GEA
890
EUR
APOSPC302899-10G
APOSPC302899-5G
APOSPC302899-1G
6-Chloro-2-fluoro-3-methoxybenzylamine 97%
6-Chloro-2-fluoro-3-methoxybenzylamine
Formula:
C₈H₉ClFNO MW: 189,62 g/mol Melting Pt: 55…57 °C Storage Temperature: Ambient |
MDL Number:
MFCD09832379 CAS Number: 1017779-65-5 UN: 3259 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...