3-Fluoro-4-(trifluoromethoxy)phenylacetic acid 98%

Supplier: Apollo Scientific
Warning

PC302956-1G PC302956-5G
APOSPC302956-1GEA 180 EUR
APOSPC302956-1G APOSPC302956-5G
3-Fluoro-4-(trifluoromethoxy)phenylacetic acid 98%
3-Fluoro-4-(trifluoromethoxy)phenylacetic acid
Formula: C₉H₆F₄O₃
MW: 238,14 g/mol
Melting Pt: 64…66 °C
Storage Temperature: Ambient
MDL Number: MFCD06660343
CAS Number: 886761-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR