4-Chloro-2-fluoro-3-methoxybenzaldehyde 98%
Supplier: Apollo Scientific
|
Warning
|
PC302973-10G
PC302973-5G
PC302973-1G
APOSPC302973-10GEA
261
EUR
APOSPC302973-10G
APOSPC302973-5G
APOSPC302973-1G
4-Chloro-2-fluoro-3-methoxybenzaldehyde 98%
4-Chloro-2-fluoro-3-methoxybenzaldehyde
Formula:
C₈H₆ClFO₂ MW: 188,58 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19687172 CAS Number: 1002344-97-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...