4-Fluoro-2-methylphenylacetonitrile 98%

Supplier: Apollo Scientific
Danger

PC302991-10G PC302991-5G PC302991-1G
APOSPC302991-10GEA 263 EUR
APOSPC302991-10G APOSPC302991-5G APOSPC302991-1G
4-Fluoro-2-methylphenylacetonitrile 98%
4-Fluoro-2-methylphenylacetonitrile
Formula: C₉H₈FN
MW: 149,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115882
CAS Number: 80141-93-1
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR