2,3,5-Trifluorobenzylamine 97%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
(2,3,5-Trifluorophenyl)methanamine
PC303012-5G
PC303012-1G
APOSPC303012-5GEA
296
EUR
APOSPC303012-5G
APOSPC303012-1G
2,3,5-Trifluorobenzylamine 97%
2,3,5-Trifluorobenzylamine
Formula:
C₇H₆F₃N MW: 161,13 g/mol Boiling Pt: 73 °C/3mmHg Storage Temperature: Ambient |
MDL Number:
MFCD00236314 CAS Number: 244022-72-8 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...