2,3-Dichloro-6-(trifluoromethyl)benzyl bromide 95%
Supplier: Apollo Scientific
Synonyms:
2-(Bromomethyl)-3,4-dichloro-1-(trifluoromethyl)benzene
PC303014-5G
PC303014-10G
PC303014-1G
PC303014-25G
APOSPC303014-5GEA
208
EUR
APOSPC303014-5G
APOSPC303014-10G
APOSPC303014-1G
APOSPC303014-25G
2,3-Dichloro-6-(trifluoromethyl)benzyl bromide 95%
2,3-Dichloro-6-(trifluoromethyl)benzyl bromide
Formula:
C₈H₄BrCl₂F₃ MW: 307.93 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06660275 CAS Number: 886501-99-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...