3-(Pentafluorothio)benzylamine

Supplier: Apollo Scientific
Danger

PC303050-10G PC303050-1G PC303050-5G
APOSPC303050-10GEA 0 EUR
APOSPC303050-10G APOSPC303050-1G APOSPC303050-5G
3-(Pentafluorothio)benzylamine
3-(Pentafluorothio)benzylamine
Formula: C₇H₈F₅NS
MW: 233,20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06213674
CAS Number: 771573-34-3
UN: 2735
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR