4-Chloro-2-(trifluoromethoxy)benzonitrile 98%

Supplier: Apollo Scientific

PC303077-10G PC303077-5G PC303077-1G
APOSPC303077-10GEA 360 EUR
APOSPC303077-10G APOSPC303077-5G APOSPC303077-1G
4-Chloro-2-(trifluoromethoxy)benzonitrile 98%
4-Chloro-2-(trifluoromethoxy)benzonitrile
Formula: C₈H₃ClF₃NO
Storage Temperature: Ambient
MDL Number: MFCD18393807
CAS Number: 1261878-30-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR