4-Ethoxy-3,5-difluoroaniline 98%

Supplier: Apollo Scientific

PC303085-10G PC303085-5G PC303085-1G
APOSPC303085-10GEA 447 EUR
APOSPC303085-10G APOSPC303085-5G APOSPC303085-1G
4-Ethoxy-3,5-difluoroaniline 98%
4-Ethoxy-3,5-difluoroaniline
Formula: C₈H₉F₂NO
MW: 173,160 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09258686
CAS Number: 942615-19-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR