6-Chloro-2-fluoro-3-methoxyaniline 98%
Supplier: Apollo Scientific
|
Warning
|
PC303100-10G
PC303100-5G
PC303100-1G
APOSPC303100-10GEA
462
EUR
APOSPC303100-10G
APOSPC303100-5G
APOSPC303100-1G
6-Chloro-2-fluoro-3-methoxyaniline 98%
6-Chloro-2-fluoro-3-methoxyaniline
Formula:
C₇H₇ClFNO Melting Pt: 40…43 °C Storage Temperature: Ambient |
MDL Number:
MFCD09832271 CAS Number: 1017777-77-3 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...