4-Methoxy-3-(trifluoromethoxy)aniline 97%

Supplier: Apollo Scientific

PC303142-10G PC303142-5G PC303142-1G
APOSPC303142-10GEA 651 EUR
APOSPC303142-10G APOSPC303142-5G APOSPC303142-1G
4-Methoxy-3-(trifluoromethoxy)aniline 97%
4-Methoxy-3-(trifluoromethoxy)aniline
Formula: C₈H₈F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD18390012
CAS Number: 647855-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR