6-Bromo-5-fluoropyridine-2-carboxylic acid

Supplier: Apollo Scientific
Warning

Synonyms: 6-Bromo-5-fluoropicolinic acid

PC305018-5G
APOSPC305018-5GEA 81.2 EUR
APOSPC305018-5G
6-Bromo-5-fluoropyridine-2-carboxylic acid
6-Bromo-5-fluoropyridine-2-carboxylic acid
Formula: C₆H₃BrFNO₂
MW: 220 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13185797
CAS Number: 1052714-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR