α,α,α,α',α',α'-Hexafluoro-3,5-xylidine 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3,5-Di(trifluoromethyl)aniline, 3,5-Bis(trifluoromethyl)aniline
PC3060-100G
PC3060-500G
PC3060-1KG
APOSPC3060-100GEA
29
EUR
APOSPC3060-100G
APOSPC3060-500G
APOSPC3060-1KG
α,α,α,α',α',α'-Hexafluoro-3,5-xylidine 98%
α,α,α,α',α',α'-Hexafluoro-3,5-xylidine
Formula:
C₈H₅F₆N MW: 229,12 g/mol Boiling Pt: 85 °C/15mmHg Density: 1,473 g/cm³ (25 °C) Flash Pt: 83 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000394 CAS Number: 328-74-5 EINECS: 206-335-0 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...