4-Aminononafluorobiphenyl 96%

Supplier: Apollo Scientific
Warning

Synonyms: Perfluoro-4-aminobiphenyl, 2,2',3,3',4',5,5',6,6'-Nonafluoro-[1,1'-biphenyl]-4-amine

PC3061-1G PC3061-250MG
APOSPC3061-1GEA 46.4 EUR
APOSPC3061-1G APOSPC3061-250MG
4-Aminononafluorobiphenyl 96%
4-Aminononafluorobiphenyl
Formula: C₁₂H₂F₉N
MW: 331.14 g/mol
Melting Pt: 145…146 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00671483
CAS Number: 969-25-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR