2-Chloro-4-methylbenzotrifluoride 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Chloro-4-methyl-1-(trifluoromethyl)benzene
PC3091-1G
PC3091-25G
PC3091-5G
APOSPC3091-1GEA
14.7
EUR
APOSPC3091-1G
APOSPC3091-25G
APOSPC3091-5G
2-Chloro-4-methylbenzotrifluoride 98%
2-Chloro-4-methylbenzotrifluoride
Formula:
C₈H₆ClF₃ MW: 194,58 g/mol Boiling Pt: 91 °C/50mmHg Flash Pt: 69 °C Storage Temperature: Ambient |
MDL Number:
MFCD00190112 CAS Number: 74483-46-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...