α,α,α,α',α',α'-Hexafluoro-5-nitroxylene 98%

Supplier: Apollo Scientific
Warning

PC3109-100G PC3109-25G PC3109-5G
APOSPC3109-100GEA 0 EUR
APOSPC3109-100G APOSPC3109-25G APOSPC3109-5G
α,α,α,α',α',α'-Hexafluoro-5-nitroxylene 98%
α,α,α,α',α',α'-Hexafluoro-5-nitroxylene
Formula: C₈H₃F₆NO₂
Boiling Pt: 72 °C/760mmHg
Density: 1,535 g/cm³ (25 °C)
Flash Pt: 77 °C
Storage Temperature: Ambient
MDL Number: MFCD00000384
CAS Number: 328-75-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR