1H,1H,11H-Perfluoroundecyl methacrylate 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1H,1H,11H-Icosafluoroundecyl methacrylate
PC3156-10G
PC3156-25G
PC3156-5G
APOSPC3156-10GEA
33.5
EUR
APOSPC3156-10G
APOSPC3156-25G
APOSPC3156-5G
1H,1H,11H-Perfluoroundecyl methacrylate 95%
1H,1H,11H-Perfluoroundecyl methacrylate
Formula:
C₁₅H₈F₂₀O₂ MW: 600,2 g/mol Boiling Pt: 110 °C/0,05mmHg Melting Pt: 36…38 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00042111 CAS Number: 41123-44-8 EINECS: 255-230-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...