Ethyl bromofluoroacetate 98%

Supplier: Apollo Scientific
Warning

Synonyms: Bromofluoroacetic ethyl ester, Ethylbromofluoroacetate

PC3164-100G PC3164-25G
APOSPC3164-100GEA 197 EUR
APOSPC3164-100G APOSPC3164-25G
Ethyl bromofluoroacetate 98%
Ethyl bromofluoroacetate

Versatile fluorinated building block. Intermediate for the synthesis of 2-fluoro-3-hydroxy esters, 2-fluoro-3-keto esters and ethyl 2-fluoroalkanoates, ask for references.

Formula: BrFCHCOOCH₂CH₃
Boiling Pt: 154 °C
Density: 1,565 g/cm³ (25 °C)
Flash Pt: 70 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00042095
CAS Number: 401-55-8
EINECS: 206-928-4
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR