1-Prop-2-ynyl-3-(trifluoromethyl)-1H-pyrazole

Supplier: Apollo Scientific
Warning

PC31707-1G PC31707-250MG
APOSPC31707-1GEA 451 EUR
APOSPC31707-1G APOSPC31707-250MG
1-Prop-2-ynyl-3-(trifluoromethyl)-1H-pyrazole
1-Prop-2-ynyl-3-(trifluoromethyl)-1H-pyrazole
Formula: C₇H₅F₃N₂
Storage Temperature: Refrigerator
MDL Number: MFCD00179866
CAS Number: 208943-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR