Ethyl-2-fluorobenzoate 98%

Supplier: Apollo Scientific
Warning

Synonyms: Ethyl 2-fluorobenzoate, 2-Fluorobenzoic acid ethyl ester

PC3210-100G PC3210-250G PC3210-25G
APOSPC3210-100GEA 20.5 EUR
APOSPC3210-100G APOSPC3210-250G APOSPC3210-25G
Ethyl-2-fluorobenzoate 98%
Ethyl 2-fluorobenzoate
Formula: C₉H₉FO₂
MW: 168,17 g/mol
Boiling Pt: 217…220 °C
Density: 1,15 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00039214
CAS Number: 443-26-5
EINECS: 207-134-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR