Ethyl-2-methyl-4,4,4-trifluorobutyrate 95%

Supplier: Apollo Scientific
Warning

PC3245K-5G PC3245K-1G
APOSPC3245K-5GEA 306 EUR
APOSPC3245K-5G APOSPC3245K-1G
Ethyl-2-methyl-4,4,4-trifluorobutyrate 95%
Ethyl-2-methyl-4,4,4-trifluorobutyrate
Formula: C₇H₁₁F₃O₂
MW: 184,16 g/mol
Boiling Pt: 142…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00190636
CAS Number: 143484-00-8
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR